Skip to Content
Merck
All Photos(1)

Documents

Z777306

Supelco

Sepax GP-C8 (5 µm) HPLC Columns

pore size 120 Å, L × I.D. 25 cm × 4.6 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

product name

Sepax GP-C8 HPLC Column, 5 μm, pore size 120 Å, L × I.D. 25 cm × 4.6 mm

material

stainless steel hardware

Agency

suitable for USP L7

feature

endcapped

packaging

pkg of 1 unit

manufacturer/tradename

Sepax 107085-4625

extent of labeling

11% Carbon loading

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4.6 mm

surface area

300 m2/g

matrix

spherical silica particle platform

matrix active group

C8 (octyl) phase

particle size

5 μm

pore size

120 Å pore size

operating pH range

2-8

separation technique

size exclusion (SEC)

General description

  • Highly controlled chemistry of monolayer formation and end-capping
  • Extremely high column-to-column reproducibility
  • High selectivity and efficiency for separations
  • Full coverage bonded silica packing to achieve the exceptionally high stability
  • Suitable for separations of acidic, neutral and basic organic compounds, such as pharmaceuticals, peptides, and organic acids

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service