Skip to Content
Merck
All Photos(1)

Documents

G005887

LC/MS (TOF) Analysis of DL-α-Hydroxyglutaric Acid Enantiomers on Astec® CHIROBIOTIC® R, Fast Separation

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Astec CHIROBIOTIC® R, 10 cm x 4.6 mm I.D., 5μm particles (13022AST)
mobile phase: 5:95, (water:methanol) with 0.3% acetic acid and 0.1% ammonium hydroxide
flow rate: 1 mL/min
pressure: 1700 psi (117 bar)
column temp.: 25 °C
detector: MS, ESI(-) TOF, SIM, m/z 147.03, split 100:1
injection: 10 μL
sample: DL-a-Hydroxyglutaric acid, 1 mg/mL in 50:50, water:methanol

suitability

application for HPLC

application(s)

clinical

Analysis Note

Chiral 2-hydroxyglutarates (2-OHG) are important molecular signatures of both healthy and diseased biological cells, their specific biochemical pathways and inborn errors of metabolism. The chiral differentiation and quantification of D-2-OHG and L-2-OHG is key for characterizing neurometabolic disorders like the 2-hydroxyglutaric acidurias that cause neurological impairment early in life. In this application, an Astec® CHIROBIOTIC® R column run under polar ionic mobile phase conditions achieved a rapid separation of the D and L enantiomers of 2-OHG under conditions amenable for sensitive LC-MS/MS detection.

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Analyte

Description

    DL-α-Hydroxyglutaric acid disodium salt, ≥95.0% (GC)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service