Key Documents
509515
SGE Gas Tight Syringes, Luer Lock
25MDR-LL-GT, volume 25 mL, needle size (not included)
Synonym(s):
SYRINGE SGE 25ML, LUER LOCK
Select a Size
1 EA
MXP 5,939.00
MXP 5,939.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
MXP 5,939.00
About This Item
UNSPSC Code:
41122001
MXP 5,939.00
Check Cart for Availability
Request a Bulk Order
material
glass syringe
needle size
(not included)
feature
needle type Luer lock
packaging
pkg of 1 ea
manufacturer/tradename
SGE 009462
volume
25 mL
General description
  • SGE gas tight syringe without a needle, Model: 25MDR-LL-GT.
  • Made of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
  • Autoclavable syringes but the plunger should be removed.
  • Features an interference fit between the plunger tip and the barrel which enhances the gas sealing properties of the syringe.
  • Maximum inertness for liquid or gas samples.
Choose from one of the most recent versions:
Certificates of Analysis (COA)