Skip to Content
Merck
All Photos(1)

Key Documents

48713

Supelco

2-Bromo-1-chloropropane solution

20,000 μg/mL in methanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6BrCl
CAS Number:
Molecular Weight:
157.44
UNSPSC Code:
12000000

grade

analytical standard

Agency

EPA 601,502.1,8021 (1987),8021A (1990)

CofA

current certificate can be downloaded

feature

standard type internal

packaging

ampule of 1 mL

concentration

20,000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C3H6BrCl/c1-3(4)2-5/h3H,2H2,1H3

InChI key

PUJJZGFFAQHYEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromo-1-chloropropane solution may be used as an analytical reference standard for the determination of the analyte in drinking water[1] and in indoor and ambient air[2] by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous determination of halogenated neutral and acidic disinfection by-products in drinking water by closed-loop stripping extraction and capillary gas chromatography.
Kampioti AA and Stephanou EG
Journal of Chromatography A, 857(1-2), 217-229 (1999)
Determination of volatile organic compounds in indoor and ambient air of residences in Kuwait.
Bouhamra WS, et al.
Environment International, 23(2), 197-204 (1997)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service