Skip to Content
Merck
All Photos(1)

Key Documents

48064

Supelco

Triphenyl phosphate solution

certified reference material, 500 μg/mL in methyl tert-butyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O)3PO
Molecular Weight:
326.28
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

certified reference material
TraceCERT®

Agency

EPA 507

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type internal

packaging

ampule of 1 mL

concentration

500 μg/mL in methyl tert-butyl ether

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

O=P(Oc1ccccc1)(Oc2ccccc2)Oc3ccccc3

InChI

1S/C18H15O4P/c19-23(20-16-10-4-1-5-11-16,21-17-12-6-2-7-13-17)22-18-14-8-3-9-15-18/h1-15H

InChI key

XZZNDPSIHUTMOC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Triphenyl phosphate may be used as an internal standard for the determination of pesticides, polycyclic aromatic hydrocarbons and polychlorinated biphenyls in environmental samples using gas chromatography coupled with mass spectrometry (GC-MS).[1]

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-phase extraction clean-up of soil and sediment extracts for the determination of various types of pollutants in a single run
Dabrowska.H, et al.
Journal of Chromatography A, 1003(1-2), 29-42 (2003)
M I Cervera et al.
Analytical and bioanalytical chemistry, 406(27), 6843-6855 (2014-05-16)
An atmospheric pressure chemical ionization source has been used to enhance the potential of gas chromatography coupled with quadrupole time-of-flight (QTOF) mass spectrometry (MS) for screening and quantification purposes in pesticide residue analysis. A screening method developed in our laboratory
Davide Savy et al.
Molecules (Basel, Switzerland), 20(11), 19958-19970 (2015-11-12)
The molecular composition of water-soluble lignins isolated from four non-food bioenergy crops (cardoon CAR, eucalyptus EUC, and two black poplars RIP and LIM) was characterized in detail, and their potential bioactivity towards maize germination and early growth evaluated. Lignins were
Johanna Jyrkkärinne et al.
Journal of medicinal chemistry, 51(22), 7181-7192 (2008-11-06)
The human constitutive androstane receptor (CAR, NR1I3) is an important regulator of xenobiotic metabolism and other physiological processes. So far, only few CAR agonists are known and no explicit mechanism has been proposed for their action. Thus, we aimed to
Boris V Krivoshiev et al.
Toxicology in vitro : an international journal published in association with BIBRA, 29(8), 2124-2132 (2015-09-08)
Flame retardants are ubiquitously used chemicals that have been shown to contaminate environments. Toxicological data is largely limited, with little insight into their molecular modes of action that may give rise to their toxic phenotypes. Such insight would aid more

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service