Skip to Content
Merck
All Photos(1)

Key Documents

47788

Supelco

EPA 552.2 Esters Calibration Mix (without Dalapon)

in methyl tert-butyl ether (varied), TraceCERT®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

analytical standard

Agency

EPA 552.2

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

in methyl tert-butyl ether (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

multi-component solution

storage temp.

2-8°C

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Methyl bromoacetate 400 μg/mL
    Methyl bromochloroacetate 400 μg/mL
    Methyl bromodichloroacetate 400 μg/mL
    Methyl chloroacetate 600 μg/mL
    Methyl chlorodibromoacetate 1000 μg/mL
    Methyl dibromoacetate 200 μg/mL
    Methyl dichloroacetate 600 μg/mL
    Methyl tribromoacetate 2000 μg/mL
    Methyl trichloroacetate 200 μg/mL
    See All (9)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service