Skip to Content
Merck
All Photos(1)

Key Documents

47130-U

Supelco

Ergosterol

10 mg/mL in chloroform, analytical standard

Synonym(s):

3β-Hydroxy-5,7,22-ergostatriene, 5,7,22-Ergostatrien-3β-ol, Provitamin D2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H44O
CAS Number:
Molecular Weight:
396.65
Beilstein:
2338604
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

10 mg/mL in chloroform

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

color

colorless

mp

156-158 °C (lit.)

application(s)

food and beverages
vitamins, nutraceuticals, and natural products

format

single component solution

functional group

hydroxyl

storage temp.

2-8°C

SMILES string

[H][C@@]1(CC[C@@]2([H])C3=CC=C4C[C@@H](O)CC[C@]4(C)[C@@]3([H])CC[C@]12C)[C@H](C)\C=C\[C@H](C)C(C)C

InChI

1S/C28H44O/c1-18(2)19(3)7-8-20(4)24-11-12-25-23-10-9-21-17-22(29)13-15-27(21,5)26(23)14-16-28(24,25)6/h7-10,18-20,22,24-26,29H,11-17H2,1-6H3/b8-7+/t19-,20+,22-,24+,25-,26-,27-,28+/m0/s1

InChI key

DNVPQKQSNYMLRS-APGDWVJJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sterols make up the majority of the unsaponifiable matter in vegetable and animal fats. Animal fats contain mostly cholesterol, whereas most vegetable fats contain only traces of this sterol. Plant sterols are collectively called phytosterols. These standards are not corrected for purity. Packed in amber ampule under nitrogen.
Ergosterol is a plant sterol known for its association with structural and growing fungal characteristics like age, maturation, hyphal formation and sporulation. It exhibits broad-spectrum health-promoting performance, by means of antioxidant, anti-inflammatory or antihyperlipidemic properties.

Application

Ergosterol may be used as an analytical reference standard for the determination of the analyte in Tuber spp. Truffles, biological samples, edible mushrooms and mouldy building materials by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification and quantification of ergosterol and phenolic compounds occurring in Tuber spp. Truffles.
Villares A, et al.
J. Food Compos. Anal., 26(1-2), 177-182 (2012)
Derivatization-independent cholesterol analysis in crude lipid extracts by liquid chromatography/mass spectrometry: applications to a rabbit model for atherosclerosis.
Shui G, et al.
Journal of Chromatography A, 1218(28), 4357-4365 (2011)
Storage temperature and UV-irradiation influence on the ergosterol content in edible mushrooms.
Villares A, et al.
Food Chemistry, 147(28), 252-256 (2014)
Determination of ergosterol on mouldy building materials using isotope dilution and gas chromatography-tandem mass spectrometry.
Nielsen KF and Madsen J?
Journal of Chromatography A, 898(2), 227-234 (2000)
Nuo Sun et al.
Antimicrobial agents and chemotherapy, 57(1), 532-542 (2012-11-14)
Mitochondrial dysfunction in pathogenic fungi or model yeast causes altered susceptibilities to antifungal drugs. Here we have characterized the role of mitochondrial complex I (CI) of Candida albicans in antifungal susceptibility. Inhibitors of CI to CV, except for CII, increased

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service