Skip to Content
Merck
All Photos(1)

Documents

23988

Supelco

Economy Moisture Trap

volume 400 cc, 1/4 in. fitting

Synonym(s):

ECONOMY MOISTURE-REMOVING TRAP, 400CC, 1/4IN FITTINGS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

volume

400 cc

fitting

1/4 in. fitting

Looking for similar products? Visit Product Comparison Guide

General description

The clear polycarbonate tubes contains a mixture of Molecular Sieve 13X and Molecular Sieve 4A with indicating capability. Because room air may permeate through the polycarbonate tube, this trap should not be used with carrier gas streams. It is designed for pneumatic control to drive equipment, such as autosamplers and valves.

The indicating 4A changes from blue to tan at 20% relative humidity.

Note: Not for use with carrier gas streams.


Dimensions:
  • 400 cc Traps: 17 1/2 in. (44.4 cm) long x 2 in. (51 mm) diameter

Application

Moisture trap may be used to remove all the solvent present, during the extraction of lipids from biological substances.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Inhalation

Target Organs

Lungs

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IMPROVED APPARATUS FOR THE EXTRACTION OF LIPIDS FROM LIQUIDS AND SOLIDS, WITH FURTHER APPLICATIONS TO THE FRACTIONATION OF FECAL FAT.
Kaye IA
The Journal of Biological Chemistry, 138 (2), 643-649 (1941)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service