20552
Premium Grade 304 Stainless Steel Tubing
tubing L × O.D. × I.D. 100 ft × 1/16 in. (1.59 mm) × 0.010 in. (0.254 mm)
Synonym(s):
SS TUBING 1/16"ODX.010ID 100FT COIL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115720
eCl@ss:
32119290
Recommended Products
material
premium grade stainless steel (304)
technique(s)
gas chromatography (GC): suitable
tubing L × O.D. × I.D.
100 ft × 1/16 in. (1.59 mm) × 0.010 in. (0.254 mm)
Looking for similar products? Visit Product Comparison Guide
General description
Grade 304 tubing, specially cleaned to ensure inertness. Use for sensitive GC-MS applications.
Type 304 stainless steel is covered by highly protective film of chromium oxyhydroxide. Hydrochloric acid solution readily attacks 304SS. Hence, it is recommended to add inhibitors like heterocyclic compounds to reduce the corrosion rate. 304SS materials are suitable for tubing and for wires as they exhibit strength and are corrosion resistant.
Application
304SS tubing may be used to make homemade reactor coil for gas chromatography oven to determine the effect of thermal volume expansion of solvents under superheated conditions.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Joseph R. Davis
Stainless Steels, 117-117 (1994)
Synthesis of Annulated Pyridines by Intramolecular Inverse-Electron-Demand Hetero-Diels-Alder Reaction under Superheated Continuous Flow Conditions.
Martin, Rainer E., et al.
European Journal of Organic Chemistry, 2012, 47-52 (2012)
Rhodanine azosulpha drugs as corrosion inhibitors for corrosion of 304 stainless steel in hydrochloric acid solution.
Abdallah, M.
Corrosion Science, 44, 717-728 (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service