T7681
Tropine 2-(4-chlorophenoxy)butanoate maleate salt
≥98%
Synonym(s):
SM21
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C18H24ClNO3 · C4H4O4
CAS Number:
Molecular Weight:
453.91
MDL number:
UNSPSC Code:
12352200
Assay
≥98%
storage temp.
2-8°C
SMILES string
OC(=O)\C=C/C(O)=O.CCC(Oc1ccc(Cl)cc1)C(=O)O[C@@H]2C[C@@H]3CC[C@H](C2)N3C
Biochem/physiol Actions
Potent analgesic. Causes increased release of acetylcholine at central muscarinic synapses.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
F Gualtieri et al.
Journal of medicinal chemistry, 37(11), 1712-1719 (1994-05-27)
Further modifications of the leads ((R)-(+)-hyoscyamine and (p-chlorophenyl)propionic acid alpha-tropanyl ester), which show analgesic and nootropic activities as a consequence of increased central presynaptic ACh release, are reported. 2-Phenoxy- and 2-(phenylthio)alkanoic acid esters showed the best results. Several members of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service