SRP6451
Fc gamma RIIIB/CD16b human
recombinant, expressed in HEK 293 cells, ≥98% (SDS-PAGE)
Synonym(s):
CD16B, FCG3, FCG3B, FCGR3, FCGR3B, IGFR3
About This Item
Recommended Products
biological source
human
recombinant
expressed in HEK 293 cells
tag
6-His tagged (C-terminus)
Assay
≥98% (SDS-PAGE)
form
lyophilized
mol wt
calculated mol wt 21.7 kDa
observed mol wt 38-50 kDa (DTT-reduced. Protein migrates due to glycosylation. Gly 17 is the predicted N-terminal.)
packaging
pkg of 10 and 50 μg
impurities
<1 EU/μg endotoxin (LAL test)
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... FCGR3B(2215)
General description
Biochem/physiol Actions
Physical form
Reconstitution
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service