Skip to Content
Merck
All Photos(1)

Key Documents

SRP6018

Sigma-Aldrich

Leptin Receptor human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

OB Protein, OBS, Obesity Protein, Obesity factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

24.5 kDa

packaging

pkg of 10 μg

impurities

Endotoxin, tested

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... LEP(3952)

General description

Leptin Binding Domain Human Recombinant also called Leptin soluble receptor produced in E.Coli is a single, non-glycosylated, polypeptide chain containing 208 amino acids and having a molecular mass of 24.5 kDa. Leptin Receptor consists of the cytokine binding domain of leptin receptor amino acids 428-635 of human leptin receptor. The Leptin Binding Domain is purified by proprietary chromatographic techniques.

Physical form

Lyophilized from 0.0045 mM NaHCO?

Reconstitution

Centrifuge the vial prior to opening. The lyophilized Leptin Binding Domain is very soluble in H?O and most aqueous buffers below and above the isoelectric point (~ 5.9).

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service