Skip to Content
Merck
All Photos(1)

Key Documents

SRP4501

Sigma-Aldrich

FABP1 human

recombinant, expressed in E. coli

Synonym(s):

FABP1, FABPL, Fatty acid-binding protein 1 liver, L-FABP, Z-protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (SDS-PAGE)

form

lyophilized

mol wt

~19 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

impurities

endotoxin, tested

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FABP1(2168)

General description

FABP1 (fatty acid-binding protein 1) belongs to the FABP family and localizes in the cytoplasm. The gene is mapped to human chromosome 2p12-q11.[1][2]
Recombinant human FABP-1 was expressed from Escherichia coli and purified through proprietary method.

Application

FABP1 human has been used in the FABP1 ELISA (enzyme-linked immunosorbent assay).[3]

Biochem/physiol Actions

FABP1 (fatty acid-binding protein 1) mainly interacts with hydrophobic lipid ligands and is involved in intracellular fatty acid transport and utilization. FABP1 interacts with the plasma membrane to help in cholesterol transfer and also allows membrane microdomain changes. It controls lipid metabolism in the liver. Mutations in the gene are associated with the risk of type 2 diabetes, non-alcoholic fatty liver disease and homeostasis assessment index (HOMA-IR). The FABP1 gene is upregulated in liver, lung, gastric and colon cancers.[1][4][2]

Physical form

Sterile filtered and Lyophilized without additives.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.
Reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liver fatty acid-binding protein (L-FABP) promotes cellular angiogenesis and migration in hepatocellular carcinoma.
Ku CY, et al.
Oncotarget, 7, 18229-18229 (2016)
Hepatitis B Virus X Protein Induces Hepatic Steatosis by Enhancing the Expression of Liver Fatty Acid Binding Protein.
Wu YL, et al.
Journal of Virology, 90, 1729-1729 (2015)
Serum liver fatty acid binding protein levels correlate positively with obesity and insulin resistance in Chinese young adults.
Shi J, et al.
PLoS ONE, 7, e48777-e48777 (2012)
Association of single-nucleotide polymorphisms rs2197076 and rs2241883 of FABP1 gene with polycystic ovary syndrome.
Xue H, et al.
Journal of Assisted Reproduction and Genetics, 33, 75-75 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service