Skip to Content
Merck
All Photos(1)

Key Documents

SRP3075

Sigma-Aldrich

IL-12p80 human

recombinant, expressed in (BTI-Tn-5B1-4) High-5 Insect Cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in (BTI-Tn-5B1-4) High-5 Insect Cells

Assay

≥95% (HPLC)
≥95% (SDS-PAGE)

form

lyophilized

mol wt

80.0 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL12B(3593)

General description

IL-12 is a disulfide-linked heterodimeric protein (p70), composed of two subunits, p35 and p40, which are encoded by two different genes. Accumulating data indicate that p40 secretion precedes that of IL-12 expression. In addition, to its ability to covalently bind to p35 to form IL-12, p40 can bind to p19 to form IL-23, or can form a homodimer designated as IL-12 p80. Elevated levels of IL-12 p80 are correlated with macrophage recruitment and increased inflammation in asthma and respiratory viral infection models. Recombinant human IL-12 p80 is an 80.0 kDa disulfide linked homodimer consisting of two p40 chains of IL-12.

Biochem/physiol Actions

IL-12 is a disulfide-linked heterodimeric protein (p70), composed of two subunits, p35 and p40, which are encoded by two different genes. Accumulating data indicate that p40 secretion precedes that of IL-12 expression. In addition, to its ability to covalently bind to p35 to form IL-12, p40 can bind to p19 to form IL-23, or can form a homodimer designated as IL-12 p80. Elevated levels of IL-12 p80 are correlated with macrophage recruitment and increased inflammation in asthma and respiratory viral infection models. Recombinant human IL-12 p80 is an 80.0 kDa disulfide linked homodimer consisting of two p40 chains of IL-12.

Sequence

p40 Subunit: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS

Physical form

Lyophilized from 10 mM Sodium Acetate, pH 5.6 + 90 mM NaCl + 2% Sucrose + 4.0% Mannitol.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Charles S Lay et al.
Nature communications, 14(1), 2882-2882 (2023-05-20)
Association of single nucleotide polymorphisms in the IL-23 receptor with several auto-inflammatory diseases, led to the heterodimeric receptor and its cytokine-ligand IL-23, becoming important drug targets. Successful antibody-based therapies directed against the cytokine have been licenced and a class of
M K Gately et al.
Journal of immunology (Baltimore, Md. : 1950), 147(3), 874-882 (1991-08-01)
IL-12 is a heterodimeric cytokine that was identified on the basis of its ability to synergize with IL-2 in the induction of cytotoxic effector cells and was originally called cytotoxic lymphocyte maturation factor (CLMF). IL-12 was also found to stimulate
M B Atkins et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 3(3), 409-417 (1997-03-01)
A Phase I dose escalation trial of i.v. administered recombinant human interleukin 12 (rhIL-12) was performed to determine its toxicity, maximum tolerated dose (MTD), pharmacokinetics, and biological and potential antineoplastic effects. Cohorts of four to six patients with advanced cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service