Skip to Content
Merck
All Photos(2)

Documents

SRP0337

Sigma-Aldrich

Tyk2 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)

Synonym(s):

JTK1, tyrosine kinase 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

38 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

0.18 mg/mL

technique(s)

activity assay: suitable
inhibition assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... TYK2(7297)

General description

Human Tyk2 (Tyrosine Kinase 2, also known as JTK1), (GenBank Accession No. NM_003331), amino acids 871-1187 (end) with N-terminal His-tag, MW=38 kDa, expressed in a Baculovirus-infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Partanen J, et al.
Proceedings of the National Academy of Sciences of the USA, 87, 8913-8917 (1991)
I Firmbach-Kraft et al.
Oncogene, 5(9), 1329-1336 (1990-09-01)
We previously identified a novel protein tyrosine kinase gene, tyk2, by screening a human lymphoid cDNA library with a tyrosine kinase domain specific c-fms restriction fragment under low stringency hybridization conditions. We have now isolated and sequenced a full length
P Domanski et al.
FEBS letters, 374(3), 317-322 (1995-11-06)
The Jak kinases and Stat transcription factors play a major role in signaling of various cytokines including IFN alpha. In this report we show a ligand-independent interaction between Tyk-2 and Jak-1 kinases. We also demonstrate that the Tyk-2 kinase forms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service