Skip to Content
Merck
All Photos(1)

Documents

SML1561

Sigma-Aldrich

LP-211

≥98% (HPLC)

Synonym(s):

6-(4-Biphenyl-2-yl-piperazin-1-yl)-hexanoic acid 4-cyano-benzylamide, N-(4-Cyanophenylmethyl)-4-(2-diphenyl)-1-piperazinehexanamide, N-[(4-Cyanophenyl)methyl]-4-[1,1′-biphenyl]-2-yl-1-piperazinehexanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H34N4O
CAS Number:
Molecular Weight:
466.62
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

film

color

colorless

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(NCC1=CC=C(C#N)C=C1)CCCCCN2CCN(C3=CC=CC=C3C4=CC=CC=C4)CC2

InChI

1S/C30H34N4O/c31-23-25-14-16-26(17-15-25)24-32-30(35)13-5-2-8-18-33-19-21-34(22-20-33)29-12-7-6-11-28(29)27-9-3-1-4-10-27/h1,3-4,6-7,9-12,14-17H,2,5,8,13,18-22,24H2,(H,32,35)

InChI key

BQEDZLDNNBDKDS-UHFFFAOYSA-N

Biochem/physiol Actions

LP-211 is a brain penetrant selective agonist for the serotonin 5-HT7 receptor with a Ki value of 0.58 nM at rat cloned 5-HT7 receptors, and >300-fold selectivity over the 5-HT1A receptor. The 5-HT7 receptor is involved in a variety of central nervous system functions including circadian rhythm, sleep, thermoregulation, nociception, and memory, and appears to be involved in cognitive disorders, anxiety, and depression. LP-211 has been used as a selective 5-HT7 receptor agonist in multiple models including studies as a potential therapeutic agent in models of Rett syndrome and Fragile X Syndrome.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service