Skip to Content
Merck
All Photos(1)

Key Documents

SCP0261

Sigma-Aldrich

H-D-Ile-Phe-Lys-pNA trifluoroacetate salt

Synonym(s):

D-Ile-Phe-Lys p-nitroanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H38N6O5
CAS Number:
Molecular Weight:
526.63
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

Assay

≥99% (TLC)

form

powder

solubility

methanol: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

CCC(C)C(N)C(=O)NC(Cc1ccccc1)C(=O)NC(CCCCN)C(=O)Nc2ccc(cc2)N(=O)=O

InChI

1S/C27H38N6O5/c1-3-18(2)24(29)27(36)32-23(17-19-9-5-4-6-10-19)26(35)31-22(11-7-8-16-28)25(34)30-20-12-14-21(15-13-20)33(37)38/h4-6,9-10,12-15,18,22-24H,3,7-8,11,16-17,28-29H2,1-2H3,(H,30,34)(H,31,35)(H,32,36)

InChI key

GPVLFJQPHXESFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

A highly specific substrate for human plasmin

Packaging

pack of 50mg and 250mg

Other Notes

Hygroscopic in nature

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service