Skip to Content
Merck
All Photos(2)

Key Documents

SAB5702858

Sigma-Aldrich

Anti-K-Cadherin (CDH6) Antibody, clone 5N2K3, Rabbit Monoclonal

Synonym(s):

CAD6, KCAD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

5N2K3, monoclonal

form

liquid

mol wt

130 kDa

species reactivity

rat, rat, human

concentration

0.2 mg/mL

technique(s)

immunoprecipitation (IP): 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

DIVPEALFLPRRTPTARDNTDVRDFINQRLKENDTDPTAPPYDSLATYAYEGTGSVADSLSSLESVTTDADQDYDYLSDWGPRFKKLADMYGGVDSDKDS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDH6(1004)

General description

This gene encodes a member of the cadherin superfamily. Cadherins are membrane glycoproteins that mediate homophilic cell-cell adhesion and play critical roles in cell differentiation and morphogenesis. The encoded protein is a type II cadherin and may play a role in kidney development as well as endometrium and placenta formation. Decreased expression of this gene may be associated with tumor growth and metastasis. [provided by RefSeq, May 2011]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 691-790 of human K-Cadherin (CDH6) (P55285).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IP

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service