Skip to Content
Merck
All Photos(2)

Documents

SAB5702242

Sigma-Aldrich

Anti-CD70 Antibody, clone 7F4S5, Rabbit Monoclonal

Synonym(s):

CD27L, CD27LG, TNFSF7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

material

colorless

clone

7F4S5, monoclonal

form

liquid

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: 1:100 - 1:800
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD70(970)

General description

CD70 is a member of the TNF family whose expression is upregulated on DCs upon maturation. Its only receptor, CD27, is expressed on T cells and NK cells. The CD70/CD27 pathway promotes effector CD8+ T cells responses (by sustaining survival of CTLs) and influences polarization of CD4+ T cells, as it represents an alternative IL-12-independent pathway of Th1 priming and suppresses effector Th17 function.

Immunogen

PBS with 0.05% proclin300,0.05% BSA,50% glycerol,pH7.3.
Recombinant protein of human CD70.

Application

WB, FC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service