Skip to Content
Merck
All Photos(1)

Key Documents

SAB5701134

Sigma-Aldrich

Anti-MonoMethyl-YAP1-K342 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
MXP 8,454.00

MXP 8,454.00


Estimated to ship onMay 13, 2025



Select a Size

Change View
100 μL
MXP 8,454.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

MXP 8,454.00


Estimated to ship onMay 13, 2025


biological source

rabbit

Quality Level

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

human, rat, mouse

concentration

0.35 mg/mL

technique(s)

western blot: 1:500-1:2000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

monomethylation (Lys342)

Gene Information

human ... YAP1(10413)

General description

This gene encodes a downstream nuclear effector of the Hippo signaling pathway which is involved in development, growth, repair, and homeostasis. This gene is known to play a role in the development and progression of multiple cancers as a transcriptional regulator of this signaling pathway and may function as a potential target for cancer treatment. Alternative splicing results in multiple transcript variants encoding different isoforms.

Immunogen

A synthetic monomethylated peptide around K342 of human YAP1 (NP_001123617.1).

Physical form

Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service