Skip to Content
Merck
All Photos(1)

Key Documents

SAB2101764

Sigma-Aldrich

Anti-PDS5A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp686B19246, Anti-FLJ41012, Anti-KIAA0648, Anti-MGC131948, Anti-PDS5, regulator of cohesion maintenance, homolog A (S. cerevisiae)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Product SAB2101764 is not currently sold in your country. Contact Technical Service

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

151 kDa

species reactivity

human, mouse, bovine, rat, rabbit, pig, horse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDS5A(23244)

Immunogen

Synthetic peptide directed towards the C terminal region of human PDS5A

Biochem/physiol Actions

PDS5A belongs to the PDS5 family. It contains 1 HEAT repeat and 3 LRR (leucine-rich) repeats. PDS5A may contribute to regulation of sister chromatid cohesion during mitosis.

Sequence

Synthetic peptide located within the following region: PPAPSKPRRGRRPKSESQGNATKNDDLNKPINKGRKRAAVGQESPGGLEA

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel Bsteh et al.
Nature communications, 14(1), 8160-8160 (2023-12-10)
Polycomb Repressive Complexes 1 and 2 (PRC1, PRC2) are conserved epigenetic regulators that promote transcriptional gene silencing. PRC1 and PRC2 converge on shared targets, catalyzing repressive histone modifications. Additionally, a subset of PRC1/PRC2 targets engage in long-range interactions whose functions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service