Skip to Content
Merck
All Photos(2)

Documents

SAB1402661

Sigma-Aldrich

Monoclonal Anti-GLYCTK antibody produced in mouse

clone 1B5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

GLYCTK1, HBEBP2, HBEBP4, HBeAgBP4A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1B5, monoclonal

form

buffered aqueous solution

mol wt

antigen ~36.63 kDa

species reactivity

mouse, human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GLYCTK(132158)

General description

This locus encodes a member of the glycerate kinase type-2 family. The encoded enzyme catalyzes the phosphorylation of (R)-glycerate and may be involved in serine degradation and fructose metabolism. Decreased activity of the encoded enzyme may be associated with the disease D-glyceric aciduria. Alternatively spliced transcript variants have been described. (provided by RefSeq)

Immunogen

GLYCTK (NP_660305.2, 332 a.a. ~ 430 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
VLSAAMQGDVKSMAQFYGLLAHVARTRLTPSMAGASVEEDAQLHELAAELQIPDLQLEEALETMAWGRGPVCLLAGGEPTVQLQGSGRGGRNQELALRV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service