Skip to Content
Merck
All Photos(1)

Key Documents

P8653

Sigma-Aldrich

Phentermine hydrochloride

Synonym(s):

α,α-Dimethylphenethylamine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N · HCl
CAS Number:
Molecular Weight:
185.69
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

drug control

USDEA Schedule IV; Home Office Schedule 3; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

SMILES string

Cl.CC(C)(N)Cc1ccccc1

InChI

1S/C10H15N.ClH/c1-10(2,11)8-9-6-4-3-5-7-9;/h3-7H,8,11H2,1-2H3;1H

InChI key

NCAIGTHBQTXTLR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Anorectic agent that is a releaser of dopamine.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Munzar et al.
Psychopharmacology, 141(3), 287-296 (1999-02-23)
To analyze the involvement of dopamine (DA) and serotonin (5-HT) release in the stimulus properties of methamphetamine, two amphetamine analogs that selectively release either brain DA (phentermine) or 5-HT (fenfluramine) were tested for their ability to substitute for methamphetamine in
N E Rowland et al.
Psychopharmacology, 149(1), 77-83 (2000-05-02)
We have shown previously that the anorectic effects of the catecholamine-releasing agent phentermine (PHEN) and the serotonin (5-HT)-releasing agent dexfenfluramine (DFEN) are greater than additive in rats. In the present study, we examined whether the norepinephrine-uptake inhibitors desmethylimipramine (DMI) and
A Balcioglu et al.
Brain research, 813(1), 67-72 (1998-11-24)
We measured the effects of acute or chronic administration of fenfluramine and phentermine, alone or in combination, on brain dopamine and serotonin release into striatal dialysates of freely moving rats. Samples collected every 30 min were assayed in a single
Eva Manakova et al.
Neuro endocrinology letters, 33 Suppl 3, 179-182 (2013-01-29)
Both, obesity as well as anorexia may be associated with infertility and other complications of pregnancy. Weight loss during pregnancy is therefore considered a risk factor. Weight loss and appetite suppressant are contraindicated during pregnancy, but the unintended exposure is
Prescrire international, 22(136), 61-64 (2013-04-18)
The cornerstones of treatment for obesity, and even more so for simple overweight, are dietary measures and physical exercise. There are no drugs with a favourable harm-benefit balance in this setting. A fixed-dose combination of topiramate, an antiepileptic drug, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service