Documents
MTCH6800200
MTC Bio Halo Lightweight Single Channel Pipettor
capacity 20-200 μL
Synonym(s):
pipette
About This Item
UNSPSC Code:
41121500
NACRES:
NB.15
packaging
pack of 1 ea
manufacturer/tradename
MTC Bio H6800-200
capacity
20-200 μL
General description
Halo Single Channel Pipettor, lightweight, 200-1250µL
Features and Benefits
  • 10-40% lighter than leading competitors
  • 4 Digit micrometer with DriftGuard protection
  • Exclusive 1250µL model for extended length tips
  • User-enabled calibration port
  • Compatible with universal tips
Legal Information
Halo is a trademark of MTC Bio
MTC is a trademark of MTC Bio