M2046
IgA, Lambda from murine myeloma
clone MOPC 315, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Mouse IgA-λ
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352203
Recommended Products
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
clone
MOPC 315, monoclonal
form
buffered aqueous solution
shipped in
dry ice
storage temp.
−20°C
Specificity
Antigen Specificity: dinitrophenyl (DNP)-modified proteins
Physical form
Solution in 0.02 M Tris buffered saline, pH 8.0, containing 0.02% sodium azide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service