GN541070
Greiner cell scrapers
handle size 25 cm
Synonym(s):
cell lifter, cell lifters, cell scraper
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122103
sterility
sterile
manufacturer/tradename
Greiner 541070
Greiner
availability
available only in China
handle size
25 cm
Looking for similar products? Visit Product Comparison Guide
General description
Greiner cell scrapers are an ideal combination of a rigid polystyrene handle with a soft polyethylene scraping blade.
Material
Handle: Polystyrene
Blade: High Density Polyethylene
Color
Handle: Blue
Blade: White
Material
Handle: Polystyrene
Blade: High Density Polyethylene
Color
Handle: Blue
Blade: White
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service