Key Documents
GE28-4064-11
Sigma-Aldrich
Tricorn 5/150 Column
About This Item
UNSPSC Code:
41121800
NACRES:
SB.52
technique(s)
LPLC: suitable
General description
Tricorn Packing Equipment includes the Packing Equipment 5/50 (5-mm connector and 50-mm glass tube) or 10/100 (10-mm connector and 100-mm glass tube), EPDM O-rings and bottom unit, cap, and stop plug.
Tricorn columns include a glass tube, adapter unit, end cap, filter kit, two stop plugs, two fingertight connectors, two M6 connectors, adapter lock, and filter holder.
Features and Benefits
Broad range of column dimensions suitable for many different purification needs.
Glass tubes coated with plastic film prevent harm if the column is dropped or operated outside its specification.
Locking ring prevents accidental compression of media bed.
Separate packing connector allows simple column packing for a broad range of media.
The modern integrated, snap-on design results in a column that is easy to assembly, operate, and maintain.
Tricorn empty columns are designed for high performance separations on small and medium scale. Pressure stability up to 100 bar and durable column material ensure reliable and reproducible operation as well as compatibility with the majority of the commercial lab media.