Skip to Content
Merck
All Photos(2)

Key Documents

GE17-0613-01

Sephacryl® S-500 HR

Cytiva 17-0613-01, pack of 750 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709
NACRES:
NA.56

packaging

pack of 750 mL

manufacturer/tradename

Cytiva 17-0613-01

matrix

allyl dextran and N, N’-methylenebisacrylamide

particle size

25-75 μm

avg. part. size

50 μm

cleaning in place

2-13

working range

3-11

suitability

suitable for bioprocess medium

General description

Sephacryl® S-500 HR is high resolution gel filtration media for preparative separation of large macromolecules, e.g. group separation of DNA restriction fragments.

Application

For preparative separation of biomolecules in the molecular weight range of 5 000 to 250 000, such as antibodies, serum proteins and mid-size proteins.

Storage and Stability

Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sephacryl is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

Sephacryl column separates using Size Exclusion Chromatography media, prepared by covalently cross-linking allyl dextran.

Column Packing and Preparation for Size Exclusion Chromatography

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service