Key Documents
F4638
Polaroid® black-and-white print film
Polaroid Type 667, This product has been discontinued. See product Z736627, Fujifilm FP-3000B, Instant Black & White Film
About This Item
UNSPSC Code:
41100000
description
Format P
ISO Daylight Speed (ASA/DIN) 3000/36°
print size 31/4 in. 41/4 in.
packaging
pack of 10 exposures
manufacturer/tradename
Polaroid Type 667
General description
Medium contrast; high speed; coaterless; professional equivalent of Type107C.
These instant films are Print Only (P) or Print-plus-Negative (P/N). The negatives require brief clearing in sodium sulfite solutions. Unless otherwise noted, prints need to be coated. Films have panchromatic spectral sensitivity. Professional equivalents have the same characteristics as their counterparts, but meet more stringent photographic specifications.
Legal Information
Polaroid is a registered trademark of Polaroid Corp.
Choose from one of the most recent versions:
Certificates of Analysis (COA)