Skip to Content
Merck
All Photos(1)

Key Documents

F1514

Sigma-Aldrich

Fmoc-L-glutamine(DOD) Sasrin® resin ester

200-400 mesh

Sign Into View Organizational & Contract Pricing

Select a Size

1 MMOL
MXP 25,862.00

MXP 25,862.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MMOL
MXP 25,862.00

About This Item

MDL number:
UNSPSC Code:
12352209
NACRES:
NA.26

MXP 25,862.00


Check Cart for Availability

Request a Bulk Order

extent of labeling

0.3-0.8 mmol FMOC-amino acid per g resin

particle size

200-400 mesh

functional group

Fmoc

storage temp.

2-8°C

SMILES string

[X]C(=O)[C@@H](NC(=O)OCC1c2c(cccc2)c3c1cccc3)CCC(=O)N

Application

For peptide synthesis. Peptides can be cleaved from the resin with 0.5-1% CF3CO2H in CH2Cl2.

Other Notes

N-FMOC-amino acid ester of 3-methoxy-4-oxymethyl-phenoxymethylated 1% divinylbenzene cross-linked polystyrene.

Legal Information

Sasrin is a registered trademark of Bachem Bioscience Inc.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service