Skip to Content
Merck
All Photos(1)

Documents

EMU202811

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Ptprc

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TAAATACTATGAAGTGTCCCTACTTGCCTATGTCAATGGGAAGATTCAAAGAAATGGGACTGCTGAGAAGTGCAATTTTCACACAAAAGCAGATCGTCCGGACAAGGTCAATGGAATGAAAACCTCCCGGCCGACAGACAATAGTATAAATGTTACATGTGGTCCTCCTTATGAAACTAATGGCCCTAAAACCTTTTACATTTTGGTAGTCAGAAGTGGAGGTTCTTTTGTTACAAAATACAACAAGACAAACTGTCAGTTTTATGTAGATAATCTCTACTATTCAACTGACTATGAGTTTCTGGTCTCTTTTCACAATGGAGTGTACGAGGGAGATTCAGTTATAAGAAATGAGTCAACAAATTTTAATGCTAAAGCACTGATTATATTCCTGGTGTTTCTGATTATTGTGACATCAATAGCCTTGCTTGTTGTTTTGTATAAAATCTATGATCTGCGCAAGAAAAGATCCAGCAATTTAGATGAACAACAGGAACTCGTTGAAAGGGA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iwona Grabowska et al.
Journal of muscle research and cell motility, 36(6), 395-404 (2015-11-29)
The skeletal muscle injury triggers the inflammatory response which is crucial for damaged muscle fiber degradation and satellite cell activation. Immunodeficient mice are often used as a model to study the myogenic potential of transplanted human stem cells. Therefore, it
Asif Manzoor Khan et al.
Neurobiology of aging, 36(6), 2164-2175 (2015-04-22)
The susceptibility of the aging brain to neurodegenerative disease may in part be attributed to cellular aging of the microglial cells that survey it. We investigated the effect of cellular aging induced by telomere shortening on microglia by the use
Xu Chang Geng et al.
Molecular medicine reports, 11(5), 3860-3865 (2015-01-13)
Erythropoietin (EPO) is a hematopoietic hormone that protects against renal interstitial fibrosis in animal models; however, the mechanism underlying the anti‑fibrotic activity of EPO has remained elusive. The present study aimed to elucidate this mechanism. Twenty‑four male C57BL6 mice were
Chiyoko Sekine et al.
Arthritis & rheumatology (Hoboken, N.J.), 66(10), 2751-2761 (2014-06-20)
We previously reported that blockade of the Notch ligand delta-like protein 1 (DLL-1) suppressed osteoclastogenesis and ameliorated arthritis in a mouse model of rheumatoid arthritis (RA). However, the mechanisms by which joint inflammation were suppressed have not yet been revealed.
Kunitoshi Kobayashi et al.
International immunology, 27(7), 333-344 (2015-02-28)
Dimethyl fumarate (DMF) is a modifier of the nuclear factor (erythroid-derived 2)-2 (Nrf2)-kelch-like ECH-associated protein 1 (Keap1) pathway. DMF treatment in the effector phase significantly suppressed the development of Theiler's murine encephalomyelitis virus-induced demyelinating disease (TMEV-IDD) both clinically and histologically.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service