Skip to Content
Merck
All Photos(1)

Key Documents

Pu-518

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
Pricing and availability is not currently available.

biological source

mouse blood

packaging

tube of 5 μg 85051501-DNA-5UG
pkg of vial of cells 85051501-1VL

growth mode

Semi-adherent

karyotype

Not specified

morphology

Macrophage

products

Not specified

receptors

Immunoglobulin, complement

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse BALB/c lymphoid macrophage

Cell Line Description

Adapted to culture from a spontaneous BALB/c mouse lymphoid tumour. Thought to be of B lymphocyte origin. Resistant to dextran sulphate. Produces isoenzyme and is capable of antibody-dependent lysis of sheep erythrocytes and tumour targets.

Culture Medium

RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Detach cells by aspirating or gentle shaking.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service