Key Documents
C8110
Culturewell coverslip system
coverslips only (5 pouches of 4 coverslips), sterile
Synonym(s):
coverslips, microscope slide, microscope slides
About This Item
UNSPSC Code:
41100000
sterility
sterile
manufacturer/tradename
Grace Bio-Labs
General description
Culturewell Coverslip System − Silicone gaskets secured to 24 × 50 mm, no. 11/2 glass coverslips. 9 mm spacing for multichannel pipettors. Leak-proof enclosures eliminate well-to-well cross contamination. Design provides convenient coverslip handling using microscope slide holders and accessories. Gaskets remove cleanly, easily for confocal microscopy and high throughput screening. Available as inserts for batch processing and as individual coverslips. Chambered Coverslip Inserts. Each package contains 10 sterile pouches, each containing 4 coverslips, and 10 sterile culture plates.
Legal Information
Culturewell is a trademark of Grace Bio-Labs, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)