C6792
Cupferron
ACS reagent
Synonym(s):
N-Nitroso-N-phenylhydroxylamine ammonium salt
About This Item
grade
ACS reagent
ign. residue
≤0.05%
color
clear
mp
150-155 °C (dec.) (lit.)
solubility
H2O: passes test
storage temp.
2-8°C
SMILES string
[H][N+]([H])([H])[H].[O-]N(N=O)c1ccccc1
InChI
1S/C6H5N2O2.H3N/c9-7-8(10)6-4-2-1-3-5-6;/h1-5H;1H3/q-1;/p+1
InChI key
GDEBSAWXIHEMNF-UHFFFAOYSA-O
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Carc. 1B - Eye Irrit. 2 - Muta. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service