Skip to Content
Merck
All Photos(1)

Key Documents

C6522

Sigma-Aldrich

Lead(II) citrate tribasic trihydrate

Synonym(s):

Citric acid lead salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5O7)2Pb3 · 3H2O
CAS Number:
Molecular Weight:
1053.85
Beilstein:
3920957
EC Number:
MDL number:
UNSPSC Code:
12171500

SMILES string

O.O.O.OC2(CC(=O)O[PbH2]OC(=O)CC1(O)CC(=O)O[PbH2]OC1=O)CC(=O)O[PbH2]OC2=O

InChI

1S/2C6H8O7.3H2O.3Pb/c2*7-3(8)1-6(13,5(11)12)2-4(9)10;;;;;;/h2*13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);3*1H2;;;/q;;;;;3*+2/p-6

InChI key

GMPMGSCJCDAUMP-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Metal stain for ultrathin sections in electron microscopy.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The use of lead citrate at high pH as an electron-opaque stain in electron microscopy.
E S REYNOLDS
The Journal of cell biology, 17, 208-212 (1963-04-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service