Key Documents
C1812
Sigma-Aldrich
Bio-transport carrier system, Nalgene®
Carrier only
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
manufacturer/tradename
Nalgene®
General description
Transparent, autoclavable system for safety transporting potentially biohazardous samples. Carrier is polycarbonate with a leakproof silicone gasket and polysulfone clamps.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)