Skip to Content
Merck
All Photos(1)

Key Documents

C167

Sigma-Aldrich

Cholera Toxin B subunit

solid

Synonym(s):

CTB

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

Quality Level

mol wt

~12 kDa

color

white

storage temp.

2-8°C

SMILES string

CCOc1ccccc1C(=O)Nc2ccc(Cl)c(c2)C(F)(F)F

InChI

1S/C16H13ClF3NO2/c1-2-23-14-6-4-3-5-11(14)15(22)21-10-7-8-13(17)12(9-10)16(18,19)20/h3-9H,2H2,1H3,(H,21,22)

InChI key

YDXZSNHARVUYNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cholera Toxin B subunit is secreted by Vibrio cholerae.[1] It is a pentamer and is nontoxic in nature.[2]

Application

Cholera Toxin B subunit has been used as a tracer to study the ascending projections of the nuclei of the descending trigeminal tract (nTTD) in the zebra finch.[3]

Biochem/physiol Actions

Cholera Toxin B subunit stimulates both systemic and mucosal antibody production in animals and humans.[2] CTB functions as an oral subunit vaccine for cholera, which is associated with acute watery diarrhoea.[1]
The cholera toxin B subunit is used for track tracing in neurological research, taking advantage of GM1 ganglioside binding and retrograde transport.
The cholera toxin B subunit is used for track tracing in neurological research, taking advantage of GM1 ganglioside binding and retrograde transport. Tissue culture cells treated with cholera toxin are not killed and tissues of animals do not become necrotic.

Caution

Do not freeze.

Reconstitution

When reconstituted in 0.25 mL distilled water, the compound contains 500 μg of protein in 0.01 M sodium phosphate at pH 7.5.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ascending projections of the nuclei of the descending trigeminal tract (nTTD) in the zebra finch (Taeniopygia guttata)
Faunes M, et al.
The Journal of Comparative Neurology, 525(5), 2832-2846 (2017)
Gangliosides as membrane receptors for tetanus toxincholera toxin and serotonin.
van Heyningen, et al.
Nature, 249, 415-417 (1974)
Efficacy of a food plant-based oral cholera toxin B subunit vaccine
Arakawa T, et al.
Nature Biotechnology, 16(3), 292-292 (1998)
J Holmgren et al.
Journal of general microbiology, 86(1), 49-65 (1975-01-01)
Structural analysis of cholera toxin by sodium dodecylsulphate polyacrylamide electrophoresis demonstrated two types of non-covalentyly linked subunits, heavy (H) AND LIGHT (L), with respective molecular weights 28000 and 800 to 9000. The H:L protein ratio was I:2, indicating that the
Karoline Lienbacher et al.
Investigative ophthalmology & visual science, 52(5), 2510-2519 (2011-01-14)
The purpose of this study was to localize the cell bodies of palisade endings that are associated with the myotendinous junctions of the extraocular muscles. Rhesus monkeys received tract-tracer injections (tetramethylrhodamine dextran [TMR-DA] or choleratoxin subunit B [CTB]) into the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service