A9654
Anti-Rat IgG (whole molecule)–Alkaline Phosphatase antibody produced in goat
affinity isolated antibody, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352203
Recommended Products
biological source
goat
conjugate
alkaline phosphatase conjugate
antibody form
affinity isolated antibody
antibody product type
secondary antibodies
clone
polyclonal
form
buffered aqueous solution
technique(s)
direct ELISA: 1:1,000
shipped in
wet ice
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Specificity
Binds all rat Igs.
Physical form
Solution in 0.05 M Tris, pH 8.0, containing 1% bovine serum albumin, 1 mM MgCl2 and 15 mM sodium azide
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Rats injected with syngenic rat apoptotic neutrophils develop antineutrophil cytoplasmic antibodies.
Yaël C Patry et al.
Journal of the American Society of Nephrology : JASN, 12(8), 1764-1768 (2001-07-20)
Antineutrophil cytoplasmic antibodies (ANCA) are present in sera from patients with various forms of vasculitis-associated glomerulonephritis. Because autoantibodies may be directed against antigens presented by apoptotic cells, generation of ANCA using apoptotic neutrophils (PMN) in syngenic Brown Norway (BN) rats
Heike Thiel et al.
Molecular plant-microbe interactions : MPMI, 22(8), 999-1010 (2009-07-11)
Beet necrotic yellow vein virus (BNYVV) induces the most important disease threatening sugar beet. The growth of partially resistant hybrids carrying monogenic dominant resistance genes stabilize yield but are unable to entirely prevent virus infection and replication. P25 is responsible
Claus-Peter Witte et al.
Plant molecular biology, 55(1), 135-147 (2004-12-18)
Beyond the rewards of plant genome analysis and gene identification, characterisation of protein activities, post-translational modifications and protein complex composition remains a challenge for plant biologists. Ideally, methods should allow rapid isolation of proteins from plant material achieving a high
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service