Skip to Content
Merck
All Photos(1)

Key Documents

46394

Sigma-Aldrich

Fluo 3-AM

suitable for fluorescence

Synonym(s):

4-(6-Acetoxymethoxy-2,7-dichloro-3-oxo-9-xanthenyl)-4′-methyl-2,2′(ethylenedioxy)dianiline-N,N,N′,N′-tetraacetic acid tetrakis(acetoxymethyl) ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H50Cl2N2O23
CAS Number:
Molecular Weight:
1129.85
UNSPSC Code:
12352200
PubChem Substance ID:

solubility

DMSO: soluble

fluorescence

λex 506 nm; λem 526 nm in 0.1 M Tris pH 8.0, esterase 30 mM Ca2+

suitability

suitable for fluorescence

storage temp.

−20°C

SMILES string

CC(=O)OCOC(=O)CN(CC(=O)OCOC(C)=O)c1ccc(C)cc1OCCOc2cc(ccc2N(CC(=O)OCOC(C)=O)CC(=O)OCOC(C)=O)C3=C4C=C(Cl)C(=O)C=C4Oc5cc(OCOC(C)=O)c(Cl)cc35

Looking for similar products? Visit Product Comparison Guide

Application

Cell permeable fluorescent indicator of intracellular Ca2+; non-fluorescent until it is hydrolyzed intracellularly and/or in the presence of Ca2+; lower binding affinity allows measurement of higher peaks of Ca2+ transients than with Fura 2.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service