Skip to Content
Merck
All Photos(1)

Key Documents

43295

Sigma-Aldrich

1-[4-(Dimethylamino)phenyl]-6-phenylhexatriene

for time-resolved fluorescence anisotropy and polarized fluorescence measurements

Synonym(s):

4′-(Dimethylamino)diphenylhexatriene, DMA-DPH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21N
CAS Number:
Molecular Weight:
275.39
Beilstein:
2940918
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥80.0% (HPLC)

form

solid

mp

198-200 °C (lit.)

fluorescence

λex 332 nm; λem 520 nm±10 nm in methanol

SMILES string

CN(C)c1ccc(\C=C\C=C\C=C\c2ccccc2)cc1

InChI

1S/C20H21N/c1-21(2)20-16-14-19(15-17-20)13-7-4-3-6-10-18-11-8-5-9-12-18/h3-17H,1-2H3/b4-3+,10-6+,13-7+

InChI key

QNBRWGUCMQGGDA-YPXUENLVSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service