Skip to Content
Merck
All Photos(1)

Key Documents

33457

Sigma-Aldrich

2-Methoxyethanol

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)

Synonym(s):

Ethylene glycol monomethyl ether, Methyl glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OCH2CH2OH
CAS Number:
Molecular Weight:
76.09
Beilstein:
1731074
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
technique(s):
GC/GC: suitable
bp:
124-125 °C (lit.)
vapor pressure:
6.17 mmHg ( 20 °C)

grade

ACS reagent
puriss. p.a.

Agency

reag. Ph. Eur.

vapor density

2.62 (vs air)

vapor pressure

6.17 mmHg ( 20 °C)

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

548 °F

expl. lim.

24.5 %

technique(s)

GC/GC: suitable

impurities

≤0.0003% peroxides (as H2O2)
≤0.001% acetaldehyde
≤0.001% formaldehyde
≤0.002% non-volatile matter
≤0.002 meq/g free acid (as CH3COOH)
≤0.1% water (Karl Fischer)

refractive index

n20/D 1.4010-1.4030
n20/D 1.402 (lit.)

bp

124-125 °C (lit.)

mp

−85 °C (lit.)

density

0.966-0.968 g/mL at 20 °C
0.965 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
B: ≤0.02 mg/kg
Ba: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Fe: ≤0.4 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sn: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

COCCO

InChI

1S/C3H8O2/c1-5-3-2-4/h4H,2-3H2,1H3

InChI key

XNWFRZJHXBZDAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methoxyethanol is widely used solvent in various electrochemical studies and industrial units. It exhibits “quasi-aprotic properties. Its solvent characteristics are a mixture of the protic and aprotic solvents. Dissolution of alkali metal salts in 2-methoxyethanol have been studied and their conductance, viscosity and laser-Raman spectra have been evaluated.[1]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B - STOT RE 2 - STOT SE 1

Target Organs

Immune system, thymus

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conductometric, viscometric, and spectroscopic investigations on the solvation phenomena of alkali-metal ions and ion pairs in 2-methoxyethanol.
Das B and Hazra DK.
The Journal of Physical Chemistry, 99(1), 269-273 (1995)
Huiping Yang et al.
Cryobiology, 61(2), 211-219 (2010-07-27)
Medaka Oryzias latipes is a well-recognized biomedical fish model because of advantageous features such as small body size, transparency of embryos, and established techniques for gene knockout and modification. The goal of this study was to evaluate two critical factors
H-Y Chang et al.
Occupational and environmental medicine, 61(8), 697-702 (2004-07-20)
To evaluate the protective effectiveness of gloves from occupational exposure to 2-methoxyethanol (2-ME); and to examine the association of 2-methoxyacetic acid (MAA) in urine and plasma collected simultaneously from low 2-ME exposure and high 2-ME exposure workers in a semiconductor
Makoto Takei et al.
Toxicological sciences : an official journal of the Society of Toxicology, 118(2), 643-652 (2010-07-10)
Ethylene glycol monomethyl ether (EGME) is a widely used industrial solvent known to cause adverse effects to human and other mammals. Organs with high metabolism and rapid cell division, such as testes, are especially sensitive to its actions. In order
Frank Welsch
Toxicology letters, 156(1), 13-28 (2005-02-12)
Numerous experimental studies have established that only a few among the large family of ethylene glycol ethers (EGEs) elicit toxicity on reproduction in either gender. Notable are the monomethyl (EGME) and monoethyl (EGEE) ethers and their respective acetate esters whose

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service