Skip to Content
Merck
All Photos(1)

Key Documents

Z800082

Hellma® liquid calibration standards

UV10, for testing the stray light

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.32

manufacturer/tradename

Hellma 667-UV10

technique(s)

UV/Vis spectroscopy: suitable

Looking for similar products? Visit Product Comparison Guide

General description

Certified liquid Hellma® calibration standard to enable the checking of spectrophotometers with respect to stray light.

Consisting of :
Sodium iodide in purified water (667-UV10, wavelength [nm] 259 (cut-off), pathlength 10 mm

Other Notes

The most current version of Hellma's handling guidelines for calibration standards can be accessed on the Hellma Analytics web site.

Legal Information

Hellma is a registered trademark of Hellma GmbH

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service