NIST1003C
Glass beads - particle size distribution
NIST® SRM® 1003c
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
powder
packaging
pkg of 28 g
manufacturer/tradename
NIST®
application(s)
glass & ceramic
Looking for similar products? Visit Product Comparison Guide
General description
This Standard Reference Material (SRM) is intended for use in the evaluation and calibration of equipment used to measure particle size distributions (PSD) in the 20 μm to 50 μm diameter range. Typical instrumentation for PSD determination would be laser light scattering (LLS), sedimentation, electrical sensing zone (ESZ) instruments, and wire-cloth test sieves in the range from No. 400 (38 μm) through No. 635 (20 μm). This size range follows that of the coarser beads of SRM 1004b. The SRM consists of a single bottle containing approximately 28 g of solid spherical soda-lime glass beads. For more information, please refer to the SDS and COA.
SRM 1003C_cert
SRM 1003C_SDS
SRM 1003C_cert
SRM 1003C_SDS
Other Notes
Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Legal Information
NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology
related product
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 1B Inhalation
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service