Skip to Content
Merck
All Photos(1)

Key Documents

F5520

Sigma-Aldrich

Fluorol Yellow 088

Synonym(s):

Solvent Green 4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16O
CAS Number:
Molecular Weight:
296.36
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

SMILES string

Cc1ccc2Oc3ccc(C)c4cc5ccccc5c(-c2c1)c34

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M C Brundrett et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 66(3), 111-116 (1991-01-01)
Polyethylene glycol (400) with 90% glycerol (aqueous) is introduced as an efficient solvent system for lipid stains. Various lipid-soluble dyes were dissolved in this solvent system and tested for their intensity, contrast, and specificity of staining of suberin lamellae in
Guilhem Reyt et al.
Current biology : CB, 30(20), 4103-4111 (2020-08-29)
Casparian strips (CSs) are cell wall modifications of vascular plants restricting extracellular free diffusion into and out of the vascular system [1]. This barrier plays a critical role in controlling the acquisition of nutrients and water necessary for normal plant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service