Skip to Content
Merck
All Photos(1)

Key Documents

C4109

Sigma-Aldrich

6-Carboxy-4′,5′-dichloro-2′,7′-dimethoxyfluorescein N-hydroxysuccinimide ester

Solid

Synonym(s):

6-JOE SE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H17Cl2NO11
CAS Number:
Molecular Weight:
602.33
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

6-Carboxy-4′,5′-dichloro-2′,7′-dimethoxyfluorescein N-hydroxysuccinimide ester,

Assay

80-99%

Quality Level

form

solid

mol wt

602.34

color

red

solubility

water: slightly soluble

λmax

(78,000 at 521 nm in 50 mmol phosphate buffer, pH 12)

fluorescence

λem 548 nm

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

wet ice

storage temp.

−20°C

SMILES string

COc1cc2c(Oc3c(Cl)c(O)c(OC)cc3C24OC(=O)c5ccc(cc45)C(=O)ON6C(=O)CCC6=O)c(Cl)c1O

InChI

1S/C27H17Cl2NO11/c1-37-15-8-13-23(19(28)21(15)33)39-24-14(9-16(38-2)22(34)20(24)29)27(13)12-7-10(3-4-11(12)26(36)40-27)25(35)41-30-17(31)5-6-18(30)32/h3-4,7-9,33-34H,5-6H2,1-2H3

InChI key

UUVBVONAGUSCCH-UHFFFAOYSA-N

General description

A traditional fluorophore used in automated DNA sequencing. Characterized by an intermediate-wavelength spectra, high quantum yield, and low pH sensitivity in the physiological range.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service