96491
Zinc ionophore I
Selectophore™, function tested
Synonym(s):
Tetrabutylthiuram disulfide
About This Item
Recommended Products
grade
for ion-selective electrodes
Quality Level
product line
Selectophore™
form
liquid
quality
function tested
SMILES string
CCCCN(CCCC)C(=S)SSC(=S)N(CCCC)CCCC
InChI
1S/C18H36N2S4/c1-5-9-13-19(14-10-6-2)17(21)23-24-18(22)20(15-11-7-3)16-12-8-4/h5-16H2,1-4H3
InChI key
PGAXJQVAHDTGBB-UHFFFAOYSA-N
General description
Application
Packaging
Other Notes
Legal Information
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service