Skip to Content
Merck
All Photos(1)

Documents

91378

Supelco

(2RS)-2-(4-Hydroxyphenyl)propionic acid

Synonym(s):

2-(4-Hydroxyphenyl)propanoic acid, 4-Hydroxyhydratropic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O3
CAS Number:
Molecular Weight:
166.17
Beilstein:
2091472
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

Assay

≥98.0% (HPLC)

Quality Level

application(s)

clinical testing

format

neat

InChI

1S/C9H10O3/c1-6(9(11)12)7-2-4-8(10)5-3-7/h2-6,10H,1H3,(H,11,12)

InChI key

ZHMMPVANGNPCBW-UHFFFAOYSA-N

Biochem/physiol Actions

2-(4-Hydroxyphenyl)propionic acid is a polyphenol metabolite detected in biological fluids. This compound belongs to the class of organic compounds known as phenylpropanoic acids. These are compounds with a structure containing a benzene ring conjugated to a propanoic acid.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Neveu et al.
Database : the journal of biological databases and curation, 2010, bap024-bap024 (2010-04-30)
A number of databases on the plant metabolome describe the chemistry and biosynthesis of plant chemicals. However, no such database is specifically focused on foods and more precisely on polyphenols, one of the major classes of phytochemicals. As antioxidants, polyphenols

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service