Skip to Content
Merck
All Photos(1)

Key Documents

62102

Supelco

Lewatit® MonoPlus SP 112 Na+ form

sodium form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

Quality Level

form

beads

parameter

120 °C max. temp. (250 °F)

moisture

53-55%

technique(s)

LPLC: suitable

loss

45-55% loss on drying, 110 °C

matrix

styrene-divinylbenzene (macroporous)

matrix active group

sulfonic acid functional group

uniform particle size

660 μm std dev ±70 μm

operating pH

0-14

capacity

1.7 meq/mL by wetted bed volume

density

1.24 g/mL at 20 °C (lit.)

bulk density

780 g/L

separation technique

cation exchange

General description

Uniform particle size (monodispersed) provides optimized kinetics and leads to an increased capacity when compared to heterodispersed bead size distribution.
spherical particles with a special particle size distribution

Application

Strongly acidic cation exchange resin especially suited for purification of condensates, demineralization of water at higher temperature, treatment of galvanic rinsing water, removal of metal ions from process solutions and passivating dips, and for the binding of heavy metals in hydrometallurgy.

Legal Information

Lewatit is a registered trademark of Lanxess Deutschland GmbH

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service