Skip to Content
Merck
All Photos(1)

Key Documents

568821

Sigma-Aldrich

Toluene

anhydrous, 99.8%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH3
CAS Number:
Molecular Weight:
92.14
Beilstein:
635760
EC Number:
MDL number:
UNSPSC Code:
12190000

grade

anhydrous

vapor density

3.2 (vs air)

vapor pressure

22 mmHg ( 20 °C)
26 mmHg ( 25 °C)

Assay

99.8%

autoignition temp.

997 °F

expl. lim.

7 %

impurities

<0.001% water

evapn. residue

<0.0005%

refractive index

n/D 1.496 (lit.)

bp

110-111 °C (lit.)

mp

-93 °C (lit.)

density

0.865 g/mL at 25 °C (lit.)

SMILES string

Cc1ccccc1

InChI

1S/C7H8/c1-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

YXFVVABEGXRONW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.9 °F - closed cup

Flash Point(C)

4.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ting Lei et al.
Small (Weinheim an der Bergstrasse, Germany), 11(24), 2946-2954 (2015-02-26)
Low-bandgap diketopyrrolopyrrole (DPP)-based polymers are used for the selective dispersion of semiconducting single-walled carbon nanotubes (s-SWCNTs). Through rational molecular design to tune the polymer-SWCNT interactions, highly selective dispersions of s-SWCNTs with diameters mainly around 1.5 nm are achieved. The influences
Liang Guo et al.
Journal of chromatography. A, 1286, 9-15 (2013-03-19)
For the first time, a novel one-step sample preparation method that combines microwave assisted extraction and solvent bar microextraction (MAE-SBME) with analysis by gas chromatography-mass spectrometry (GC-MS), was developed for the fast and efficient determination of polycyclic aromatic hydrocarbons (PAHs)
Elke Persch et al.
ChemMedChem, 9(8), 1880-1891 (2014-05-03)
The causative agents of the parasitic disease human African trypanosomiasis belong to the family of trypanosomatids. These parasitic protozoa exhibit a unique thiol redox metabolism that is based on the flavoenzyme trypanothione reductase (TR). TR was identified as a potential
Tayseer Mahdi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(31), 11134-11142 (2015-06-26)
Frustrated Lewis pairs (FLPs) based on sterically encumbered anilines and the Lewis acid B(C6 F5 )3 were found to react with terminal alkynes effecting intermolecular hydroamination affording iminium alkynylborate species of the form [RPhNC(R')Me][R'CCB(C6 F5 )3 ]. In these cases
Yoko Yamakoshi et al.
Faraday discussions, 173, 287-296 (2014-12-04)
The photoinduced reactive oxygen species (ROS) generation from several water-soluble fullerenes was examined. Macromolecular or small molecular water-soluble fullerene complexes/derivatives were prepared and their (1)O(2) and O(2)˙(-) generation abilities were evaluated by ESR spin-trapping methods. As a result, efficient (1)O(2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service