Skip to Content
Merck
All Photos(1)

Key Documents

55951

Supelco

Etoxazole

reference material

Synonym(s):

(RS)-5-tert-Butyl-2-[2-(2,6-difluorophenyl)-4,5-dihydro-1,3-oxazol-4-yl]phenetole, 4-(4-tert-Butyl-2-ethoxyphenyl)-2-(2,6-difluorophenyl)-4,5-dihydrooxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23F2NO2
CAS Number:
Molecular Weight:
359.41
Beilstein:
8930214
MDL number:
UNSPSC Code:
41116107

grade

reference material

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

format

neat

SMILES string

CCOc1cc(ccc1C2COC(=N2)c3c(F)cccc3F)C(C)(C)C

InChI

1S/C21H23F2NO2/c1-5-25-18-11-13(21(2,3)4)9-10-14(18)17-12-26-20(24-17)19-15(22)7-6-8-16(19)23/h6-11,17H,5,12H2,1-4H3

InChI key

IXSZQYVWNJNRAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO/IEC 17025. This RM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

854.6 °F

Flash Point(C)

457 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wonhyoung Park et al.
Environmental pollution (Barking, Essex : 1987), 257, 113480-113480 (2019-11-21)
Etoxazole is an organofluorine insecticide widely used in agriculture. Exposure to insecticides is a serious environmental problem owing to their cytotoxic effects in humans and animals. Reproductive toxicity of various organofluorine insecticides have been shown in previous studies. However, few

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service